Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AACS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319839.1 | NP_001306768.1 | 4, 5 | 681 | ||
NM_001319840.1 | NP_001306769.1 | 4, 5 | 681 | |||
NM_023928.4 | NP_076417.2 | 4, 5 | 681 | |||
XM_011538692.2 | XP_011536994.1 | 2, 3 | 192 | |||
GenBank mRNA | AB054121.1 | - | 4, 5 | 570 | ||
AK022451.1 | - | 4, 5 | 611 | |||
AK022740.1 | - | 2, 3 | 221 | |||
AK024036.1 | - | 4, 5 | 610 | |||
AK304176.1 | - | 5, 6 | 883 | |||
AK308779.1 | - | 5, 6 | 889 | |||
AL833047.1 | - | 3, 4 | 322 | |||
AY091468.1 | - | 4, 5 | 442 | |||
BC040490.1 | - | 4, 5 | 669 | 5295926 | ||
BC041000.1 | - | 4, 5 | 664 | 5273267 | ||
BC051862.1 | - | 4, 5 | 587 | 5762683 |