Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDRGK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023935.2 | NP_076424.1 | 4 | 515 | ||
GenBank mRNA | AK310809.1 | - | 4 | 498 | ||
BC000643.1 | - | 4 | 467 | 3344932 | ||
BC007957.1 | - | 4 | 473 | 4301923 | ||
BC011851.1 | - | 4 | 468 | 4136696 |