Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYNX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023946.3 | NP_076435.1 | 3 | 382 | ||
NM_177457.4 | NP_803252.1 | 3 | 382 | |||
NM_177476.3 | NP_803429.1 | 3 | 401 | |||
NM_177477.3 | NP_803430.1 | 3 | 515 | |||
GenBank mRNA | AF321824.1 | - | 3 | 341 | ||
BC014560.2 | - | 2 | 568 | |||
BG701653.1 | - | 3 | 400 | |||
BI603715.1 | - | 3 | 519 | |||
BM172253.1 | - | 3 | 400 | |||
BM671222.1 | - | 4 | 423 | |||
BQ685632.1 | - | 3 | 330 | |||
HY110957.1 | - | 3 | 399 |