Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYNX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023946.3 | NP_076435.1 | 4 | 492 | ||
NM_177458.1 | NP_803253.1 | 2 | 158 | |||
GenBank mRNA | AB081838.1 | - | 2 | 158 | ||
AY358417.1 | - | 2 | 174 | |||
AY587277.1 | - | 2 | 136 | |||
BM671222.1 | - | 2 | 290 |