Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142674.1 | NP_001136146.1 | 7 | 702 | ||
NM_001142675.1 | NP_001136147.1 | 6 | 812 | |||
NM_001142676.1 | NP_001136148.1 | 7 | 673 | |||
NM_001142677.1 | NP_001136149.1 | 6 | 598 | |||
NM_023947.3 | NP_076436.3 | 6 | 598 | |||
XM_005253072.2 | XP_005253129.1 | 6 | 685 | |||
XM_005253073.4 | XP_005253130.1 | 7 | 686 | |||
XM_006718282.2 | XP_006718345.1 | 7 | 673 | |||
XM_011520299.1 | XP_011518601.1 | 6 | 909 | |||
XM_011520300.1 | XP_011518602.1 | 6 | 909 | |||
XM_011520301.2 | XP_011518603.1 | 6 | 909 | |||
XM_011520302.2 | XP_011518604.1 | 6 | 909 | |||
XM_017018166.1 | XP_016873655.1 | 7 | 759 | |||
XM_017018167.1 | XP_016873656.1 | 6 | 564 | |||
XM_017018168.1 | XP_016873657.1 | 7 | 685 | |||
GenBank mRNA | AF212229.1 | - | 7 | 636 | ||
AK027711.1 | - | 6 | 547 | |||
AK075413.1 | - | 7 | 609 | |||
AK124697.1 | - | 6 | 1174 | |||
AK225025.1 | - | 7 | 638 | |||
AK225398.1 | - | 7 | 621 | |||
AK301245.1 | - | 6 | 658 | |||
AY037151.1 | - | 6 | 559 | |||
BC000001.2 | - | 7 | 622 | 3504261 | ||
BC013642.1 | - | 4 | 234 | |||
BC095409.1 | - | 6 | 534 | 30333427 | ||
BI818660.1 | - | 7 | 702 | |||
BI821281.1 | - | 6 | 564 |