Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003076.4 | NP_003067.3 | 8 | 1125 | ||
NM_139071.2 | NP_620710.2 | 8 | 1125 | |||
XM_005269107.3 | XP_005269164.2 | 7 | 1002 | |||
GenBank mRNA | AF109733.1 | - | 8 | 913 | ||
AK130517.1 | - | 9 | 865 | |||
AK292700.1 | - | 8 | 789 | |||
BC009368.2 | - | 8 | 1075 | 4110499 | ||
CN272379.1 | - | 7 | 655 | |||
DA825057.1 | - | 4 | 396 | |||
U66617.1 | - | 8 | 1103 |