Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024938.3 | NP_001020109.1 | 12 | 1726 | ||
NM_001024939.3 | NP_001020110.1 | 12 | 1614 | |||
NM_001282864.1 | NP_001269793.1 | 11 | 1968 | |||
NM_030807.4 | NP_110434.3 | 13 | 1837 | |||
NR_104247.1 | 11 | 1573 | ||||
GenBank mRNA | AB209301.1 | - | 8 | 2199 | ||
AF443201.1 | - | 13 | 1668 | |||
AJ271290.1 | - | 12 | 1557 | |||
AK075467.1 | - | 12 | 1502 | |||
AK092466.1 | - | 2 | 1638 | |||
AK300196.1 | - | 11 | 1967 | |||
AK314502.1 | - | 13 | 1608 | |||
BC018967.2 | - | 1 | 282 | |||
BC094735.1 | - | 12 | 1543 | 5304025 | ||
BC100807.2 | - | 9 | 1675 | |||
BC100808.2 | - | 12 | 1472 | 40000757 | ||
BC100809.1 | - | 12 | 1472 | 40000760 | ||
BC100810.1 | - | 11 | 1319 | |||
CR456373.1 | - | 12 | 1503 |