Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003093.2 | NP_003084.1 | 6 | 915 | ||
NR_029472.1 | 5 | 872 | ||||
GenBank mRNA | AI356808.1 | - | 1 | 298 | ||
AK223098.1 | - | 6 | 489 | |||
AK311955.1 | - | 6 | 495 | |||
BC121082.1 | - | 6 | 484 | 40119776 | ||
BC121083.1 | - | 6 | 484 | 40119777 | ||
BU153661.1 | - | 5 | 411 | |||
BU508721.1 | - | 5 | 441 | |||
M18465.1 | - | 4 | 389 | |||
X12517.1 | - | 6 | 480 |