Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174044.1 | NP_001167515.1 | 3 | 1038 | ||
NM_001174045.1 | NP_001167516.1 | 4 | 1033 | |||
NM_001174046.1 | NP_001167517.1 | 3 | 948 | |||
NM_003117.4 | NP_003108.2 | 3 | 859 | |||
NM_153189.2 | NP_694859.1 | 3 | 859 | |||
GenBank mRNA | AK292229.1 | - | 3 | 948 | ||
AY920278.1 | - | 3 | 1034 | |||
AY920279.1 | - | 3 | 855 | |||
AY920280.1 | - | 4 | 1008 | |||
AY920281.1 | - | 3 | 923 | |||
BC026163.1 | - | 3 | 782 | 4838230 | ||
EU794686.1 | - | 3 | 822 | |||
L13781.1 | - | 1 | 521 | |||
S67798.1 | - | 3 | 793 | |||
X84347.1 | - | 2 | 716 |