Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPTBN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003128.2 | NP_003119.2 | 17 | 3862 | ||
NM_178313.2 | NP_842565.2 | 16 | 3959 | |||
XM_005264517.2 | XP_005264574.1 | 17 | 3771 | |||
XM_005264518.1 | XP_005264575.1 | 16 | 4617 | |||
XM_006712087.2 | XP_006712150.1 | 17 | 3735 | |||
XM_017004779.1 | XP_016860268.1 | 17 | 3878 | |||
XM_017004780.1 | XP_016860269.1 | 17 | 3832 | |||
XM_017004781.1 | XP_016860270.1 | 17 | 3833 | |||
GenBank mRNA | AB209748.1 | - | 17 | 3740 | ||
AB371586.1 | - | 17 | 3852 | |||
AF327441.1 | - | 16 | 3959 | |||
BC137282.1 | - | 16 | 3869 | 9021279 | ||
BC137283.1 | - | 16 | 3869 | 9021280 | ||
EU794662.1 | - | 17 | 3862 | |||
JQ599728.1 | - | 16 | 3613 | |||
S65762.1 | - | 9 | 2737 |