Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001725.2 | NP_001716.2 | 13, 14 | 1406 | ||
GenBank mRNA | AF322588.1 | - | 13, 14 | 1332 | ||
AK296568.1 | - | 13, 14 | 1251 | |||
AK315328.1 | - | 13, 14 | 1373 | |||
AX713278.1 | - | 13, 14 | 1374 | |||
AX746123.1 | - | 13, 14 | 1374 | |||
AX752831.1 | - | 13, 14 | 1374 | |||
BC040955.1 | - | 13, 14 | 1381 | 5218655 | ||
DQ414688.1 | - | 13, 14 | 1344 | |||
J04739.1 | - | 13, 14 | 1374 |