Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SREBF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004599.3 | NP_004590.2 | 13 | 2627 | ||
NR_103834.1 | 14 | 2727 | ||||
XM_006724310.2 | XP_006724373.1 | 13 | 2871 | |||
XM_011530347.1 | XP_011528649.1 | 11 | 2081 | |||
XM_017028921.1 | XP_016884410.1 | 13 | 2456 | |||
GenBank mRNA | AK299914.1 | - | 13 | 2456 | ||
AL834221.1 | - | 5 | 986 | |||
BC051385.1 | - | 14 | 2633 | 5497929 | ||
BC051799.1 | - | 13 | 2593 | |||
BC056158.1 | - | 13 | 2573 | 6169568 | ||
CT841522.1 | - | 13 | 2517 | |||
U02031.1 | - | 13 | 2552 |