Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001309434.1 | NP_001296363.1 | 3 | 258 | ||
NM_003134.5 | NP_003125.3 | 3 | 258 | |||
GenBank mRNA | AB451391.1 | - | 3 | 172 | ||
BC035495.2 | - | 3 | 214 | 4298087 | ||
BC071716.1 | - | 3 | 194 | 4476669 | ||
BC100031.1 | - | 3 | 198 | 2847600 | ||
U07857.1 | - | 3 | 193 | |||
X73459.1 | - | 3 | 191 |
Gene Symbol: SMCO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011542910.2 | XP_011541212.1 | 1 | 338 | ||
XM_017018019.1 | XP_016873508.1 | 1 | 335 |