Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007294.3 | NP_009225.1 | 23 | 5744 | ||
NM_007297.3 | NP_009228.2 | 22 | 5652 | |||
NM_007298.3 | NP_009229.2 | 22 | 2219 | |||
NM_007299.3 | NP_009230.2 | 22 | 2320 | |||
NM_007300.3 | NP_009231.2 | 24 | 5807 | |||
NR_027676.1 | 23 | 5648 | ||||
GenBank mRNA | AF005068.1 | - | 23 | 5613 | ||
BC072418.1 | - | 22 | 2320 | |||
BC115037.1 | - | 24 | 5712 | 7961446 | ||
DQ333386.1 | - | 8 | 586 | |||
DQ333387.1 | - | 5 | 368 | |||
DQ363751.1 | - | 11 | 1087 | |||
JN686490.1 | - | 22 | 5512 | |||
KP272106.1 | - | 3 | 147 | |||
U14680.1 | - | 23 | 5631 | |||
U64805.1 | - | 22 | 2200 | |||
U68041.1 | - | 9 | 622 |