Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRP72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267722.1 | NP_001254651.1 | 13, 14 | 1354 | ||
NM_006947.3 | NP_008878.3 | 15, 16 | 1537 | |||
XM_005265765.4 | XP_005265822.3 | 15, 16 | 2256 | |||
GenBank mRNA | AF038851.1 | - | 15, 16 | 1509 | ||
AF069765.1 | - | 15, 16 | 1507 | |||
AF077019.1 | - | 15, 16 | 1497 | |||
AK225430.1 | - | 15, 16 | 1500 | |||
AK307397.1 | - | 15, 16 | 1500 | |||
BC017057.1 | - | 15, 16 | 1525 | |||
BC032609.1 | - | 15, 16 | 1515 | |||
BC040134.1 | - | 15, 16 | 1518 | |||
BC046143.1 | - | 15, 16 | 1509 | |||
BC065018.1 | - | 15, 16 | 1509 | |||
BC105583.1 | - | 15, 16 | 1525 | |||
BX537991.1 | - | 13, 14 | 1334 | |||
EU794663.1 | - | 15, 16 | 1507 |