Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292008.1 | NP_001278937.1 | 8 | 837 | ||
NM_003144.4 | NP_003135.2 | 8 | 1041 | |||
NR_120448.1 | 7 | 962 | ||||
GenBank mRNA | AF156965.1 | - | 8 | 958 | ||
AK075562.1 | - | 7 | 825 | |||
AK222762.1 | - | 8 | 909 | |||
AK222809.1 | - | 8 | 909 | |||
AK291550.1 | - | 8 | 909 | |||
AK312653.1 | - | 8 | 919 | |||
AL550910.3 | - | 8 | 698 | |||
BC007710.2 | - | 8 | 934 | 4054116 | ||
CR936719.1 | - | 7 | 900 | |||
Z12830.1 | - | 8 | 876 |