Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292008.1 | NP_001278937.1 | 4, 5 | 529 | ||
NM_003144.4 | NP_003135.2 | 4, 5 | 733 | |||
NR_120448.1 | 4, 5 | 733 | ||||
GenBank mRNA | AF156965.1 | - | 4, 5 | 650 | ||
AK075562.1 | - | 3, 4 | 517 | |||
AK222762.1 | - | 4, 5 | 601 | |||
AK222809.1 | - | 4, 5 | 601 | |||
AK291550.1 | - | 4, 5 | 601 | |||
AK312653.1 | - | 4, 5 | 611 | |||
AL550910.3 | - | 4, 5 | 390 | |||
BC007710.2 | - | 4, 5 | 626 | 4054116 | ||
BQ216448.1 | - | 4, 5 | 523 | |||
BT007387.1 | - | 4, 5 | 539 | |||
CR456961.1 | - | 4, 5 | 539 | |||
CR936719.1 | - | 3, 4 | 592 | |||
Z12830.1 | - | 4, 5 | 568 |