Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005419.3 | NP_005410.1 | 6 | 718 | ||
NM_198332.1 | NP_938146.1 | 6 | 706 | |||
XM_005269111.3 | XP_005269168.2 | 6 | 626 | |||
XM_011538697.2 | XP_011536999.1 | 6 | 626 | |||
XM_011538698.2 | XP_011537000.1 | 6 | 587 | |||
XM_011538699.2 | XP_011537001.1 | 6 | 626 | |||
XM_017019904.1 | XP_016875393.1 | 6 | 626 | |||
GenBank mRNA | AK295052.1 | - | 6 | 606 | ||
AK295114.1 | - | 6 | 473 | |||
AK296939.1 | - | 6 | 579 | |||
AK296940.1 | - | 6 | 579 | |||
AK310727.1 | - | 5 | 735 | |||
AK316516.1 | - | 6 | 602 | |||
BC006092.1 | - | 5 | 505 | |||
BC051284.1 | - | 5 | 538 | 6460486 | ||
M97934.1 | - | 6 | 552 |