Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243835.1 | NP_001230764.1 | 7 | 869 | ||
NM_003151.3 | NP_003142.1 | 7 | 920 | |||
XM_006712719.3 | XP_006712782.1 | 7 | 915 | |||
XM_011511705.2 | XP_011510007.1 | 10 | 1066 | |||
XM_017004784.1 | XP_016860273.1 | 7 | 674 | |||
GenBank mRNA | AK299976.1 | - | 7 | 641 | ||
AK300868.1 | - | 7 | 673 | |||
AK301172.1 | - | 5 | 344 | |||
BC031212.1 | - | 7 | 841 | 4830583 | ||
L78440.1 | - | 7 | 686 |