Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STXBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007269.2 | NP_009200.2 | 9 | 852 | ||
GenBank mRNA | AF032922.1 | - | 9 | 834 | ||
AK223170.1 | - | 9 | 831 | |||
AK290134.1 | - | 9 | 836 | |||
AK291322.1 | - | 9 | 832 | |||
AK312910.1 | - | 9 | 829 | |||
BC038099.2 | - | 9 | 791 | 4826366 | ||
BC047764.1 | - | 9 | 827 | 5585614 | ||
D63506.1 | - | 9 | 828 |