Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SURF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278942.1 | NP_001265871.1 | 4, 5 | 674 | ||
NM_006753.5 | NP_006744.2 | 4, 5 | 675 | |||
NR_103874.1 | 4, 5 | 622 | ||||
GenBank mRNA | AK054884.1 | - | 4, 5 | 672 | ||
BC003001.1 | - | 4, 5 | 602 | |||
BC006197.2 | - | 4, 5 | 602 | 2820773 | ||
BC014878.1 | - | 4, 5 | 637 | 4343122 | ||
BI820270.1 | - | 4, 5 | 617 | |||
CN260811.1 | - | 4, 5 | 561 |