Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUV39H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282166.1 | NP_001269095.1 | 3 | 726 | ||
NM_003173.3 | NP_003164.1 | 3 | 479 | |||
GenBank mRNA | AF019968.1 | - | 3 | 472 | ||
AK223071.1 | - | 3 | 451 | |||
AK297566.1 | - | 758 | ||||
AK299900.1 | - | 3 | 726 | |||
AK312547.1 | - | 3 | 451 | |||
BC006238.2 | - | 3 | 470 | 3945632 | ||
CR541746.1 | - | 3 | 427 |