Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293725.1 | NP_001280654.1 | 8, 9 | 1790 | ||
NM_005640.2 | NP_005631.1 | 8, 9 | 1775 | |||
NR_121653.1 | 8, 9 | 1775 | ||||
XM_005258339.3 | XP_005258396.1 | 8, 9 | 2167 | |||
XM_011526153.2 | XP_011524455.1 | 8, 9 | 2167 | |||
XM_017025932.1 | XP_016881421.1 | 8, 9 | 2167 | |||
XM_017025933.1 | XP_016881422.1 | 8, 9 | 1399 | |||
GenBank mRNA | AB234096.1 | - | 8, 9 | 2187 | ||
AK302593.1 | - | 7, 8 | 1264 | |||
AY888048.1 | - | 8, 9 | 1775 | |||
AY888049.1 | - | 8, 9 | 1775 | |||
Y09321.1 | - | 8, 9 | 1541 |