Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015892.1 | NP_001015892.1 | 3 | 561 | ||
NM_003187.4 | NP_003178.1 | 3 | 324 | |||
GenBank mRNA | AI879232.1 | - | 3 | 281 | ||
BC003400.1 | - | 3 | 252 | 3448836 | ||
BC033320.1 | - | 3 | 1112 | 4837792 | ||
BG490028.1 | - | 3 | 384 | |||
BG720149.1 | - | 3 | 304 | |||
BT019652.1 | - | 1 | 94 | |||
CX784292.1 | - | 3 | 363 | |||
U21858.1 | - | 3 | 241 | |||
U25112.1 | - | 2 | 140 | |||
U30504.1 | - | 2 | 140 |