Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015892.1 | NP_001015892.1 | 3 | 883 | ||
NM_003187.4 | NP_003178.1 | 3 | 646 | |||
GenBank mRNA | BC003400.1 | - | 3 | 574 | 3448836 | |
BC033320.1 | - | 3 | 1434 | 4837792 | ||
BG720149.1 | - | 3 | 626 | |||
BT019652.1 | - | 1 | 416 | |||
U21858.1 | - | 3 | 563 | |||
U25112.1 | - | 2 | 462 | |||
U30504.1 | - | 2 | 462 |