Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000545.6 | NP_000536.5 | 1 | 267 | ||
NM_001306179.1 | NP_001293108.1 | 1 | 267 | |||
XM_005253931.3 | XP_005253988.1 | 1 | 320 | |||
GenBank mRNA | BC143483.1 | - | 1 | 86 | ||
HM116541.1 | - | 1 | 209 | |||
HM116542.1 | - | 1 | 209 | |||
HM116543.1 | - | 1 | 209 | |||
HM116544.1 | - | 1 | 209 | |||
HM116547.1 | - | 1 | 209 | |||
HM116548.1 | - | 1 | 209 | |||
HM116557.1 | - | 1 | 184 | |||
HM116558.1 | - | 1 | 184 | |||
HM449088.1 | - | 1 | 89 | |||
HM449089.1 | - | 1 | 89 | |||
M57732.1 | - | 1 | 89 | |||
X71346.1 | - | 1 | 262 | |||
X71347.1 | - | 1 | 198 |