Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093728.2 | NP_001087197.1 | 9 | 1389 | ||
NM_001261817.1 | NP_001248746.1 | 9 | 1374 | |||
NM_001261818.1 | NP_001248747.1 | 8 | 1251 | |||
NM_001261819.1 | NP_001248748.1 | 8 | 1236 | |||
NM_001261820.1 | NP_001248749.1 | 10 | 1595 | |||
NM_001261821.1 | NP_001248750.1 | 10 | 1580 | |||
NM_018679.5 | NP_061149.1 | 9 | 1483 | |||
XM_005249339.2 | XP_005249396.1 | 8 | 1144 | |||
XM_011514830.1 | XP_011513132.1 | 9 | 1621 | |||
XM_011514831.1 | XP_011513133.1 | 7 | 1057 | |||
XM_011514832.1 | XP_011513134.1 | 6 | 921 | |||
GenBank mRNA | AF260330.1 | - | 6 | 932 | ||
AF269223.1 | - | 8 | 1167 | |||
AF536532.1 | - | 10 | 1435 | |||
AF536533.1 | - | 9 | 1422 | |||
AK057234.1 | - | 9 | 1206 | |||
AK098590.1 | - | 7 | 978 | |||
AK301778.1 | - | 9 | 1265 | |||
AK301975.1 | - | 8 | 1172 | |||
AK302022.1 | - | 8 | 1137 | |||
AK302375.1 | - | 9 | 1314 | |||
AK302456.1 | - | 9 | 1295 | |||
AK315078.1 | - | 9 | 1416 | |||
AK316403.1 | - | 9 | 1198 | |||
AY069943.1 | - | 8 | 1191 | |||
BC033729.1 | - | 9 | 1335 | 5167179 | ||
BC040003.1 | - | 10 | 1437 | 5741923 | ||
BC048418.1 | - | 9 | 1413 | 5298037 | ||
BC090050.1 | - | 10 | 1441 | 30387768 |