Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022171.2 | NP_071503.1 | 1 | 411 | ||
GenBank mRNA | AK000824.1 | - | 1 | 228 | ||
AK311839.1 | - | 1 | 194 | |||
BC005157.1 | - | 1 | 377 | 2959904 | ||
CR457411.1 | - | 1 | 190 | |||
L41143.1 | - | 1 | 411 |