Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNLT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006520.2 | NP_006511.1 | 5 | 440 | ||
GenBank mRNA | AK311851.1 | - | 5 | 375 | ||
BC000968.2 | - | 5 | 368 | 3448623 | ||
CR450320.1 | - | 5 | 314 | |||
CR542213.1 | - | 5 | 314 | |||
DA342302.1 | - | 5 | 431 | |||
DB492712.1 | - | 5 | 438 | |||
U02556.1 | - | 5 | 382 |