Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291602.1 | NP_001278531.1 | 2 | 157 | ||
NM_001291603.1 | NP_001278532.1 | 3 | 199 | |||
NM_006519.3 | NP_006510.1 | 3 | 199 | |||
GenBank mRNA | AK026669.1 | - | 2 | 1525 | ||
AK315601.1 | - | 3 | 156 | |||
BC029412.1 | - | 3 | 152 | 4074224 | ||
BC105588.1 | - | 3 | 148 | 6723966 | ||
BU675683.1 | - | 3 | 586 | |||
CR456931.1 | - | 3 | 121 | |||
D50663.1 | - | 3 | 133 | |||
DN915418.1 | - | 2 | 113 | |||
EU862237.1 | - | 3 | 125 | |||
HY017024.1 | - | 2 | 110 | |||
HY171700.1 | - | 3 | 202 | |||
U56255.1 | - | 3 | 121 |