Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145398.2 | NP_001138870.1 | 4 | 840 | ||
NM_003216.3 | NP_003207.1 | 4 | 846 | |||
GenBank mRNA | AK091916.1 | - | 4 | 718 | ||
AK295595.1 | - | 4 | 772 | |||
AK314906.1 | - | 4 | 762 | |||
BC039258.1 | - | 4 | 815 | 5286096 | ||
BC042476.2 | - | 4 | 816 | 4915095 | ||
BX537848.1 | - | 2 | 243 | |||
CR456592.1 | - | 4 | 743 | |||
CR541827.1 | - | 4 | 730 | |||
U06935.1 | - | 4 | 654 | |||
U44059.1 | - | 4 | 766 |