Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099691.2 | NP_001093161.1 | 1, 2 | 281 | ||
NM_003236.3 | NP_003227.1 | 1, 2 | 281 | |||
GenBank mRNA | AF149096.1 | - | 1, 2 | 38 | ||
AF149097.1 | - | 1, 2 | 38 | |||
AF149098.1 | - | 1, 2 | 38 | |||
AK290151.1 | - | 1, 2 | 92 | |||
AK312899.1 | - | 1, 2 | 92 | |||
BC005308.1 | - | 1, 2 | 92 | 3828239 | ||
BT006833.1 | - | 1, 2 | 33 | |||
DA216480.1 | - | 1, 2 | 283 | |||
DA462271.1 | - | 1, 2 | 274 | |||
K03222.1 | - | 1, 2 | 67 | |||
M31172.1 | - | 1, 2 | 67 | |||
X70340.1 | - | 1, 2 | 64 |