Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGFB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003239.3 | NP_003230.1 | 5 | 1951 | ||
GenBank mRNA | AK122902.1 | - | 2 | 1203 | ||
AK313564.1 | - | 5 | 1032 | |||
BC018503.1 | - | 5 | 922 | 4477175 | ||
BT007287.1 | - | 5 | 822 | |||
EF560714.1 | - | 5 | 935 | |||
J03241.1 | - | 5 | 1084 | |||
X14149.1 | - | 5 | 1075 |