Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGFBI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000358.2 | NP_000349.1 | 10 | 1457 | ||
GenBank mRNA | AB209598.1 | - | 4 | 4219 | ||
AK093916.1 | - | 10 | 1312 | |||
AK094055.1 | - | 11 | 1388 | |||
AK094581.1 | - | 10 | 1401 | |||
AK222833.1 | - | 10 | 1365 | |||
AK298603.1 | - | 5 | 677 | |||
AK303643.1 | - | 10 | 1689 | |||
BC000097.2 | - | 10 | 1350 | 3506051 | ||
BC004972.1 | - | 10 | 1345 | 2958878 | ||
BT009820.1 | - | 10 | 1296 | |||
KU178357.1 | - | 10 | 1296 | |||
KU178358.1 | - | 8 | 810 | |||
M77349.1 | - | 10 | 1343 |