Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THRSP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003251.3 | NP_003242.1 | 1 | 418 | ||
GenBank mRNA | AK311975.1 | - | 1 | 418 | ||
BC031989.1 | - | 1 | 401 | 4745757 | ||
BG674197.1 | - | 1 | 371 | |||
BP317892.1 | - | 1 | 418 | |||
CB122257.1 | - | 1 | 418 |