Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000714.5 | NP_000705.2 | 4 | 569 | ||
NM_001256530.1 | NP_001243459.1 | 4 | 764 | |||
NM_001256531.1 | NP_001243460.1 | 4 | 673 | |||
NR_046308.1 | 3 | 358 | ||||
GenBank mRNA | AF075589.1 | - | 3 | 372 | ||
AF075590.1 | - | 3 | 372 | |||
AY998017.1 | - | 3 | 464 | |||
BG749252.1 | - | 4 | 673 | |||
BG752086.1 | - | 4 | 570 | |||
CR456409.1 | - | 3 | 458 |