Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003253.2 | NP_003244.2 | 5 | 953 | ||
XM_005261037.1 | XP_005261094.1 | 4 | 1086 | |||
XM_005261038.2 | XP_005261095.1 | 3 | 820 | |||
XM_005261040.2 | XP_005261097.1 | 4 | 1086 | |||
XM_011529711.1 | XP_011528013.1 | 5 | 1037 | |||
XM_011529712.1 | XP_011528014.1 | 5 | 961 | |||
XM_011529713.1 | XP_011528015.1 | 5 | 1371 | |||
XM_017028447.1 | XP_016883936.1 | 5 | 1140 | |||
XM_017028448.1 | XP_016883937.1 | 5 | 1110 | |||
XM_017028449.1 | XP_016883938.1 | 4 | 1017 | |||
XM_017028450.1 | XP_016883939.1 | 4 | 1037 | |||
XM_017028451.1 | XP_016883940.1 | 4 | 1517 | |||
XM_017028452.1 | XP_016883941.1 | 6 | 1317 | |||
XM_017028453.1 | XP_016883942.1 | 4 | 992 | |||
GenBank mRNA | BC117192.1 | - | 2 | 528 | 40125743 | |
BC117196.1 | - | 2 | 528 | 40125747 | ||
BC143970.1 | - | 2 | 528 | |||
BC143980.1 | - | 2 | 528 | 9052498 | ||
U16296.1 | - | 5 | 954 |