Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253357.1 | NP_001240286.1 | 4 | 660 | ||
NM_005424.4 | NP_005415.1 | 4 | 743 | |||
XM_005271163.1 | XP_005271220.1 | 4 | 686 | |||
XM_006710869.1 | XP_006710932.1 | 4 | 877 | |||
XM_017002207.1 | XP_016857696.1 | 4 | 686 | |||
GenBank mRNA | AK225560.1 | - | 4 | 621 | ||
AK300399.1 | - | 4 | 550 | |||
AK300623.1 | - | 5 | 572 | |||
BC038239.1 | - | 4 | 645 | 5767075 | ||
EU826586.1 | - | 4 | 566 | |||
EU826587.1 | - | 4 | 566 | |||
EU826588.1 | - | 4 | 566 | |||
EU826590.1 | - | 4 | 566 | |||
X60957.1 | - | 4 | 602 |