Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000362.4 | NP_000353.1 | 2, 3 | 1392 | ||
GenBank mRNA | AK314871.1 | - | 2, 3 | 501 | ||
BC014277.2 | - | 2, 3 | 449 | 3689386 | ||
BT006848.1 | - | 2, 3 | 206 | |||
CR456593.1 | - | 2, 3 | 254 | |||
L15078.1 | - | 2, 3 | 164 | |||
S78453.1 | - | 2, 3 | 482 | |||
U02571.1 | - | 2, 3 | 487 | |||
U14394.1 | - | 2, 3 | 493 | |||
U67195.1 | - | 2, 3 | 484 | |||
X76227.1 | - | 2, 3 | 276 |