Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003256.3 | NP_003247.1 | 4, 5 | 978 | ||
GenBank mRNA | AK313018.1 | - | 4, 5 | 539 | ||
BC010553.1 | - | 4, 5 | 978 | 4154090 | ||
BT019627.1 | - | 4, 5 | 467 | |||
U76456.1 | - | 4, 5 | 526 |