Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003258.4 | NP_003249.3 | 7 | 767 | ||
XM_017024992.1 | XP_016880481.1 | 6 | 870 | |||
GenBank mRNA | AB451268.1 | - | 7 | 557 | ||
AB451395.1 | - | 7 | 557 | |||
AK314950.1 | - | 7 | 610 | |||
BC006484.1 | - | 7 | 597 | 2905608 | ||
BC007872.2 | - | 7 | 584 | 4303880 | ||
BC007986.2 | - | 7 | 593 | 2966331 | ||
BT006941.1 | - | 7 | 557 | |||
K02581.1 | - | 7 | 614 |