Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004616.2 | NP_004607.1 | 6, 7 | 625 | ||
XM_006719583.3 | XP_006719646.1 | 6, 7 | 579 | |||
XM_017019913.1 | XP_016875402.1 | 5, 6 | 661 | |||
GenBank mRNA | BC005246.1 | - | 6, 7 | 625 | 3934330 | |
BC070168.1 | - | 10, 11 | 999 | |||
M35252.1 | - | 5, 6 | 583 |