Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278740.1 | NP_001265669.1 | 5 | 919 | ||
NM_001278741.1 | NP_001265670.1 | 5 | 689 | |||
NM_001278742.1 | NP_001265671.1 | 5 | 689 | |||
NM_001278743.1 | NP_001265672.1 | 5 | 689 | |||
NM_003270.3 | NP_003261.1 | 5 | 627 | |||
XM_011531018.1 | XP_011529320.1 | 6 | 429 | |||
GenBank mRNA | AB097016.1 | - | 5 | 523 | ||
AB209876.1 | - | 5 | 586 | |||
AF043906.1 | - | 5 | 531 | |||
AF053453.1 | - | 5 | 483 | |||
AF133426.1 | - | 5 | 490 | |||
AK075072.1 | - | 5 | 576 | |||
AK299176.1 | - | 6 | 346 | |||
AK300852.1 | - | 6 | 567 | |||
AK309581.1 | - | 5 | 902 | |||
AK315773.1 | - | 5 | 609 | |||
AY358825.1 | - | 5 | 504 | |||
BC012389.1 | - | 5 | 567 | 3857537 | ||
BT006977.1 | - | 5 | 464 | |||
CN274931.1 | - | 5 | 409 | |||
CR457115.1 | - | 5 | 464 | |||
CX760536.1 | - | 5 | 627 | |||
CX868833.1 | - | 5 | 398 | |||
DA038474.1 | - | 5 | 427 | |||
U84895.1 | - | 5 | 531 |