Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003290.2 | NP_003281.1 | 1, 2 | 371 | ||
XM_005260042.3 | XP_005260099.1 | 1, 2 | 153 | |||
GenBank mRNA | AF186109.1 | - | 1, 2 | 133 | ||
AK300136.1 | - | 1, 2 | 229 | |||
AK301285.1 | - | 1, 2 | 201 | |||
AK315076.1 | - | 1, 2 | 175 | |||
BC002827.2 | - | 1, 2 | 178 | 3637300 | ||
BC037576.1 | - | 1, 2 | 246 | 5582453 | ||
BC067225.1 | - | 1, 2 | 200 | 4772340 | ||
BT019634.1 | - | 1, 2 | 130 | |||
DC365500.1 | - | 1, 2 | 372 | |||
FJ224305.1 | - | 1, 2 | 180 | |||
X05276.1 | - | 1, 2 | 180 |