Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286272.1 | NP_001273201.1 | 1, 2 | 239 | ||
NM_003295.3 | NP_003286.1 | 1, 2 | 239 | |||
GenBank mRNA | AF072098.1 | - | 119 | |||
AK130847.1 | - | 1, 2 | 138 | |||
AK296587.1 | - | 1, 2 | 138 | |||
AK296952.1 | - | 1, 2 | 138 | |||
AK312951.1 | - | 1, 2 | 138 | |||
AY117678.1 | - | 1, 2 | 139 | |||
AY334563.1 | - | 1, 2 | 24 | |||
BC003352.2 | - | 1, 2 | 122 | 2899964 | ||
BC012431.1 | - | 1, 2 | 98 | 3880559 | ||
BC052333.1 | - | 1, 2 | 128 | 6058799 | ||
BP215975.1 | - | 1, 2 | 239 | |||
CR457036.1 | - | 1, 2 | 24 | |||
X16064.1 | - | 1, 2 | 118 | |||
X64899.1 | - | 1, 2 | 111 |