Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286272.1 | NP_001273201.1 | 3, 4 | 518 | ||
NM_001286273.1 | NP_001273202.1 | 2, 3 | 444 | |||
NM_003295.3 | NP_003286.1 | 3, 4 | 518 | |||
GenBank mRNA | AK130847.1 | - | 3, 4 | 417 | ||
AK312951.1 | - | 3, 4 | 417 | |||
AY117678.1 | - | 3, 4 | 418 | |||
AY334563.1 | - | 3, 4 | 303 | |||
BC003352.2 | - | 3, 4 | 401 | 2899964 | ||
BC012431.1 | - | 3, 4 | 377 | 3880559 | ||
BC022436.1 | - | 2, 3 | 372 | 4281364 | ||
BC052333.1 | - | 3, 4 | 407 | 6058799 | ||
BP215975.1 | - | 3, 4 | 519 | |||
CR457036.1 | - | 3, 4 | 303 | |||
L13806.1 | - | 1, 2 | 138 | |||
X16064.1 | - | 3, 4 | 397 | |||
X64899.1 | - | 3, 4 | 389 |