Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021138.3 | NP_066961.2 | 5 | 539 | ||
XM_011518973.2 | XP_011517275.1 | 6 | 843 | |||
XM_011518974.2 | XP_011517276.1 | 5 | 1444 | |||
XM_011518975.2 | XP_011517277.1 | 6 | 771 | |||
XM_011518976.2 | XP_011517278.1 | 6 | 812 | |||
XM_011518977.2 | XP_011517279.1 | 5 | 532 | |||
XM_011518978.2 | XP_011517280.1 | 5 | 1953 | |||
XM_017015094.1 | XP_016870583.1 | 5 | 587 | |||
XM_017015095.1 | XP_016870584.1 | 5 | 1297 | |||
GenBank mRNA | AK054686.1 | - | 5 | 682 | ||
AK222961.1 | - | 5 | 536 | |||
AK289722.1 | - | 6 | 655 | |||
AK298370.1 | - | 5 | 503 | |||
BC032410.1 | - | 4 | 525 | 5216790 | ||
BC033810.1 | - | 5 | 527 | 5182942 | ||
BC043492.1 | - | 5 | 539 | 5767794 | ||
BC064662.1 | - | 4 | 534 | 6528158 | ||
BM785733.1 | - | 5 | 539 | |||
BX538160.1 | - | 5 | 556 | |||
HM991672.1 | - | 4 | 482 |