Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000064.3 | NP_000055.2 | 21 | 2817 | ||
GenBank mRNA | AK304071.1 | - | 19 | 2368 | ||
BC150179.1 | - | 21 | 2756 | 40148091 | ||
BC150200.1 | - | 21 | 2756 | 40148812 | ||
BP201387.1 | - | 3 | 346 | |||
EU794602.1 | - | 21 | 2756 | |||
K02765.1 | - | 21 | 2784 |