Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPYL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003309.3 | NP_003300.1 | 1 | 1341 | ||
GenBank mRNA | AK026849.1 | - | 1 | 202 | ||
AL136629.1 | - | 1 | 1155 | |||
BC048969.1 | - | 1 | 1157 | 5297724 | ||
BC068554.1 | - | 1 | 1154 | |||
CR533535.1 | - | 1 | 1057 |