Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4BPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000716.3 | NP_000707.1 | 6 | 956 | ||
NM_001017364.1 | NP_001017364.1 | 6 | 953 | |||
NM_001017365.2 | NP_001017365.1 | 7 | 818 | |||
NM_001017366.2 | NP_001017366.1 | 7 | 815 | |||
NM_001017367.1 | NP_001017367.1 | 7 | 792 | |||
XM_005273254.4 | XP_005273311.1 | 8 | 1097 | |||
GenBank mRNA | AK092427.1 | - | 3 | 1842 | ||
AK315137.1 | - | 7 | 1058 | |||
BC005378.1 | - | 6 | 953 | 3934453 | ||
BM987840.1 | - | 1 | 172 | |||
L11245.1 | - | 6 | 941 | |||
M29964.1 | - | 6 | 691 |